Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.2315 in, 2.0000 in OAL
Catalog No. 5000537571
Encompass
This Class ZZ Plus Plug Gage features a diameter of 0.2315 in and an overall length of 2.0000 in. Manufactured from tool steel, it offers a tolerance of 0.000200 in and a 10 Micro finish. It is suitable for Go Inspection and for setting various measuring devices, and includes a Certificate of Accuracy with NIST Traceability.
  • Includes certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Suitable for Go inspection and setting various measuring devices.
Catalog No. 50-005-37571 Supplier MSC Supplier No. 88923156
This Class ZZ Plus Plug Gage features a diameter of 0.2315 in and an overall length of 2.0000 in. Manufactured from tool steel, it offers a tolerance of 0.000200 in and a 10 Micro finish. It is suitable for Go Inspection and for setting various measuring devices, and includes a Certificate of Accuracy with NIST Traceability.
  • Includes certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Suitable for Go inspection and setting various measuring devices.