Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.2340 in, 2.0000 in OAL, Tool Steel
Catalog No. 5000530753
Encompass
This plug gage is designed for precise measurement applications, including go inspection and setting various measuring devices. It features tool steel construction and includes a certificate of accuracy with NIST traceability.
  • Has a tolerance of 0.000200 in.
  • Features a 10 micro coating/finish.
  • Constructed from durable tool steel.
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
Catalog No. 50-005-30753 Supplier MSC Supplier No. 88923404
This plug gage is designed for precise measurement applications, including go inspection and setting various measuring devices. It features tool steel construction and includes a certificate of accuracy with NIST traceability.
  • Has a tolerance of 0.000200 in.
  • Features a 10 micro coating/finish.
  • Constructed from durable tool steel.
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.