Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.2670 in Diameter, 2 in Length
Catalog No. 5000531457
Encompass
This Class ZZ Plus Plug Gage is engineered for precision go inspection. It features a 0.2670 in diameter, a 2.0000 in overall length, and is crafted from tool steel with a 10 microinch finish. With a tolerance of 0.000200 in, this gage includes a Certificate of Accuracy with NIST Traceability and complies with ASME B89.1 standards.
  • Designed for go inspection, setting micrometers, verniers, and other measuring devices.
  • Made from durable tool steel.
  • Features a 10 microinch finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Diameter of measurement: 0.2670 in.
  • Overall length: 2.0000 in.
  • Tolerance: 0.000200 in.
Catalog No. 50-005-31457 Supplier MSC Supplier No. 88926704
This Class ZZ Plus Plug Gage is engineered for precision go inspection. It features a 0.2670 in diameter, a 2.0000 in overall length, and is crafted from tool steel with a 10 microinch finish. With a tolerance of 0.000200 in, this gage includes a Certificate of Accuracy with NIST Traceability and complies with ASME B89.1 standards.
  • Designed for go inspection, setting micrometers, verniers, and other measuring devices.
  • Made from durable tool steel.
  • Features a 10 microinch finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Diameter of measurement: 0.2670 in.
  • Overall length: 2.0000 in.
  • Tolerance: 0.000200 in.