Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.2690 in Diameter, Tool Steel
Catalog No. 5000527383
Encompass
This plug gage provides precise measurement and inspection, featuring a Class ZZ Plus tolerance for high accuracy. Made from durable tool steel, it is suitable for 'go' inspection and calibrating various measuring devices.
  • Class ZZ Plus tolerance for accuracy
  • 0.2690 inch diameter
  • 2.0000 inch overall length
  • 0.000200 inch tolerance
  • Tool steel construction
  • 10 micro finish
  • Meets ASME B89.1 standards
  • For 'go' inspection and setting measuring devices
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-27383 Supplier MSC Supplier No. 88926902
This plug gage provides precise measurement and inspection, featuring a Class ZZ Plus tolerance for high accuracy. Made from durable tool steel, it is suitable for 'go' inspection and calibrating various measuring devices.
  • Class ZZ Plus tolerance for accuracy
  • 0.2690 inch diameter
  • 2.0000 inch overall length
  • 0.000200 inch tolerance
  • Tool steel construction
  • 10 micro finish
  • Meets ASME B89.1 standards
  • For 'go' inspection and setting measuring devices
  • Includes certificate of accuracy with NIST traceability