Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.2720 in Diameter, 2.0000 in OAL
Catalog No. 5000527385
Encompass
This plug gage is designed for precise measurement and inspection of hole diameters. Featuring a Class ZZ Plus tolerance, it provides high accuracy for quality control applications. The gage has a diameter of 0.2720 in and an overall length of 2.0000 in.
  • Precise measurement of hole diameters.
  • High accuracy with Class ZZ Plus tolerance.
  • Durable construction for extended use.
  • Suitable for quality control and inspection tasks.
Catalog No. 50-005-27385 Supplier MSC Supplier No. 88927207
This plug gage is designed for precise measurement and inspection of hole diameters. Featuring a Class ZZ Plus tolerance, it provides high accuracy for quality control applications. The gage has a diameter of 0.2720 in and an overall length of 2.0000 in.
  • Precise measurement of hole diameters.
  • High accuracy with Class ZZ Plus tolerance.
  • Durable construction for extended use.
  • Suitable for quality control and inspection tasks.