Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.2915 in Diameter, Tool Steel
Catalog No. 5000535747
Encompass
Vermont Gage Class ZZ Plus Plug Gages are designed for precise measurement applications, including hole diameters, distances between holes, slot widths, and locations. They are also suitable for Go/NoGo inspection and for setting micrometers, verniers, and other measuring devices.
  • Made from durable tool steel
  • Features a 0.0002 in tolerance
  • Provides a 10 microinch finish for accuracy
  • Measures 2.0000 in overall length
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Ideal for Go inspection and setting measuring devices
Catalog No. 50-005-35747 Supplier MSC Supplier No. 88929153
Vermont Gage Class ZZ Plus Plug Gages are designed for precise measurement applications, including hole diameters, distances between holes, slot widths, and locations. They are also suitable for Go/NoGo inspection and for setting micrometers, verniers, and other measuring devices.
  • Made from durable tool steel
  • Features a 0.0002 in tolerance
  • Provides a 10 microinch finish for accuracy
  • Measures 2.0000 in overall length
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Ideal for Go inspection and setting measuring devices