Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.3105 in Diameter, 2.0000 in Overall Length
Catalog No. 5000515733
Encompass
This Class ZZ Plus plug gage offers precise measurement capabilities. It is manufactured with specific dimensions, including its diameter, overall length, and tolerance, ensuring accuracy for critical applications. The gage also features a fine microfinish for consistent and reliable performance.
  • Class ZZ Plus for high precision
  • 0.3105 inch diameter
  • 2.0000 inch overall length
  • 0.000200 inch tolerance
  • 10 microfinish
Catalog No. 50-005-15733 Supplier MSC Supplier No. 88931050
This Class ZZ Plus plug gage offers precise measurement capabilities. It is manufactured with specific dimensions, including its diameter, overall length, and tolerance, ensuring accuracy for critical applications. The gage also features a fine microfinish for consistent and reliable performance.
  • Class ZZ Plus for high precision
  • 0.3105 inch diameter
  • 2.0000 inch overall length
  • 0.000200 inch tolerance
  • 10 microfinish