Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.3250 in, 2.0000 in OAL
Catalog No. 5000515396
Encompass
This Class ZZ Plus Plug Gage is engineered from durable tool steel for precise measurement applications. With a 0.3250 inch diameter and 2.0000 inch overall length, it is ideal for go inspection and setting various measuring devices such as micrometers and verniers. Each gage includes a Certificate of Accuracy with NIST traceability, ensuring reliable and compliant performance.
  • Engineered from durable tool steel
  • 0.3250 in diameter for precise measurements
  • 2.0000 in overall length
  • Features a tight tolerance of 0.000200 in
  • Ideal for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • 10 micro coating/finish for enhanced durability
Catalog No. 50-005-15396 Supplier MSC Supplier No. 88932504
This Class ZZ Plus Plug Gage is engineered from durable tool steel for precise measurement applications. With a 0.3250 inch diameter and 2.0000 inch overall length, it is ideal for go inspection and setting various measuring devices such as micrometers and verniers. Each gage includes a Certificate of Accuracy with NIST traceability, ensuring reliable and compliant performance.
  • Engineered from durable tool steel
  • 0.3250 in diameter for precise measurements
  • 2.0000 in overall length
  • Features a tight tolerance of 0.000200 in
  • Ideal for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • 10 micro coating/finish for enhanced durability