Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.37 mm Diameter, 2.0000 in OAL
Catalog No. 5000528603
Encompass
This plug gage is designed for precise 'Go' inspection and for setting various measuring devices such as micrometers and verniers. Constructed from durable tool steel with a black oxide finish, it offers reliable performance. Each gage meets ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability, ensuring high precision for critical measurements.
  • Certificate of accuracy with NIST traceability included
  • Constructed from tool steel
  • Black oxide finish
  • Meets ASME B89.1 standards
  • Suitable for go inspection
  • Ideal for setting micrometers, verniers, and other measuring devices
Catalog No. 50-005-28603 Supplier MSC Supplier No. 50795640
This plug gage is designed for precise 'Go' inspection and for setting various measuring devices such as micrometers and verniers. Constructed from durable tool steel with a black oxide finish, it offers reliable performance. Each gage meets ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability, ensuring high precision for critical measurements.
  • Certificate of accuracy with NIST traceability included
  • Constructed from tool steel
  • Black oxide finish
  • Meets ASME B89.1 standards
  • Suitable for go inspection
  • Ideal for setting micrometers, verniers, and other measuring devices