Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.3920 in Diameter, 0.000200 in Tolerance
Catalog No. 5000513793
Encompass
The Class ZZ Plus Plug Gage has a 0.3920 in diameter and an overall length of 2.0000 in. It features a tolerance of 0.000200 in and a 10 Micro coating/finish. Constructed from tool steel, this plug gage is suitable for various measurement tasks.
  • Designed for go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Certificate of accuracy with NIST traceability included
Catalog No. 50-005-13793 Supplier MSC Supplier No. 88939202
The Class ZZ Plus Plug Gage has a 0.3920 in diameter and an overall length of 2.0000 in. It features a tolerance of 0.000200 in and a 10 Micro coating/finish. Constructed from tool steel, this plug gage is suitable for various measurement tasks.
  • Designed for go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Certificate of accuracy with NIST traceability included