Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.4310 in, 2.0000 in, Tool Steel
Catalog No. 5000531969
Encompass
This Class ZZ Plus Plug Gage is designed for precise 'Go' inspection, setting micrometers, verniers, and other measuring devices. It features a 0.4310 in diameter and an overall length of 2.0000 in, crafted from durable tool steel.
  • Includes certificate of accuracy with NIST traceability.
  • Product offers a hassle-free guarantee for exchange or refund within 30 days.
  • Made from tool steel.
  • Features a precision tolerance of 0.000200 in.
  • Has a 10 micro coating/finish.
  • Meets ASME B89.1 standards.
Catalog No. 50-005-31969 Supplier MSC Supplier No. 88943105
This Class ZZ Plus Plug Gage is designed for precise 'Go' inspection, setting micrometers, verniers, and other measuring devices. It features a 0.4310 in diameter and an overall length of 2.0000 in, crafted from durable tool steel.
  • Includes certificate of accuracy with NIST traceability.
  • Product offers a hassle-free guarantee for exchange or refund within 30 days.
  • Made from tool steel.
  • Features a precision tolerance of 0.000200 in.
  • Has a 10 micro coating/finish.
  • Meets ASME B89.1 standards.