Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.4535 in Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000535770
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Made from durable tool steel, it features a 0.4535 inch diameter and a 2.0000 inch overall length, ensuring accuracy with a tight tolerance of 0.000200 "
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured to ASME B89.1 standards.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-35770 Supplier MSC Supplier No. 88945357
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Made from durable tool steel, it features a 0.4535 inch diameter and a 2.0000 inch overall length, ensuring accuracy with a tight tolerance of 0.000200 "
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured to ASME B89.1 standards.
  • Features a 10 micro coating/finish.