Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.4540 in Dia, 2.0000 in OAL
Catalog No. 5000511333
Encompass
Vermont Gage Class ZZ Plus Plug Gages are designed for precise go inspection and for setting micrometers, verniers, and other measuring devices. These gages feature a diameter of 0.4540 inch and an overall length of 2.0000 inch, crafted from durable tool steel with a 10 micro finish. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable measurements.
  • Includes certificate of accuracy with NIST traceability
  • Application for go inspection, setting micrometers, verniers, and other measuring devices
  • Manufactured from tool steel
  • Features a 10 micro finish
  • Meets ASME B89.1 standards
  • Diameter of measurement: 0.4540 in
  • Overall length: 2.0000 in
  • Tolerance: 0.000200 in
Catalog No. 50-005-11333 Supplier MSC Supplier No. 88945407
Vermont Gage Class ZZ Plus Plug Gages are designed for precise go inspection and for setting micrometers, verniers, and other measuring devices. These gages feature a diameter of 0.4540 inch and an overall length of 2.0000 inch, crafted from durable tool steel with a 10 micro finish. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable measurements.
  • Includes certificate of accuracy with NIST traceability
  • Application for go inspection, setting micrometers, verniers, and other measuring devices
  • Manufactured from tool steel
  • Features a 10 micro finish
  • Meets ASME B89.1 standards
  • Diameter of measurement: 0.4540 in
  • Overall length: 2.0000 in
  • Tolerance: 0.000200 in