Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.4755 in Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000515642
Encompass
This Class ZZ Plus Plug Gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.4755 in diameter and a 2.0000 in overall length, manufactured from durable tool steel with a tolerance of 0.000200 in.
  • Designed for Go inspection and setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from durable tool steel.
  • Features a 10 micro finish.
  • Meets ASME B89.1 standards.
Catalog No. 50-005-15642 Supplier MSC Supplier No. 88947551
This Class ZZ Plus Plug Gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.4755 in diameter and a 2.0000 in overall length, manufactured from durable tool steel with a tolerance of 0.000200 in.
  • Designed for Go inspection and setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from durable tool steel.
  • Features a 10 micro finish.
  • Meets ASME B89.1 standards.