Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.5340 in Diameter, 2.0000 in Length
Catalog No. 5000513836
Encompass
This precision measuring tool is designed for accurately inspecting hole sizes and depths. Manufactured from durable tool steel, it offers exceptional wear resistance and reliability for quality control applications.
  • Precisely measures hole diameters.
  • Features a 0.5340 in diameter.
  • Offers an overall length of 2.0000 in.
  • Conforms to Class ZZ tolerance of +0.0002/-0.0000 in.
  • Has a 10 microinch surface finish or better for smooth operation.
  • Constructed from hardened tool steel.
Catalog No. 50-005-13836 Supplier MSC Supplier No. 88953401
This precision measuring tool is designed for accurately inspecting hole sizes and depths. Manufactured from durable tool steel, it offers exceptional wear resistance and reliability for quality control applications.
  • Precisely measures hole diameters.
  • Features a 0.5340 in diameter.
  • Offers an overall length of 2.0000 in.
  • Conforms to Class ZZ tolerance of +0.0002/-0.0000 in.
  • Has a 10 microinch surface finish or better for smooth operation.
  • Constructed from hardened tool steel.