Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.5740 in Diameter
Catalog No. 5000530877
Encompass
This Class ZZ Plus plug gage is designed for precision measurement. It features a 0.5740 inch diameter, 2.0000 inch overall length, and a tolerance of 0.000200 inch, making it suitable for 'Go' inspection, and for setting micrometers, verniers, and other measuring devices. Crafted from tool steel, it comes with a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards.
  • 0.5740 in diameter
  • 2.0000 in overall length
  • 0.000200 in tolerance
  • Constructed from tool steel
  • Ideal for "go" inspection and setting measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
  • Features a 10 micro coating/finish
Catalog No. 50-005-30877 Supplier MSC Supplier No. 88957402
This Class ZZ Plus plug gage is designed for precision measurement. It features a 0.5740 inch diameter, 2.0000 inch overall length, and a tolerance of 0.000200 inch, making it suitable for 'Go' inspection, and for setting micrometers, verniers, and other measuring devices. Crafted from tool steel, it comes with a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards.
  • 0.5740 in diameter
  • 2.0000 in overall length
  • 0.000200 in tolerance
  • Constructed from tool steel
  • Ideal for "go" inspection and setting measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
  • Features a 10 micro coating/finish