Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.6000 in Dia, 2.0000 in OAL
Catalog No. 5000531573
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate Go Inspection and for calibrating various measuring devices. Made from durable tool steel, it features a 0.6000 in diameter, a 2.0000 in overall length, a tight tolerance of 0.000200 in, and a smooth 10 Micro finish.
  • Designed for go inspection.
  • Used for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
Catalog No. 50-005-31573 Supplier MSC Supplier No. 88960000
The Vermont Gage Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate Go Inspection and for calibrating various measuring devices. Made from durable tool steel, it features a 0.6000 in diameter, a 2.0000 in overall length, a tight tolerance of 0.000200 in, and a smooth 10 Micro finish.
  • Designed for go inspection.
  • Used for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.