Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.7050 in Diameter, 0.000200 in Tolerance
Catalog No. 5000515096
Encompass
This Class ZZ Plus plug gage is designed for precision inspection and calibration tasks. It features a 0.000200 in tolerance and a 10 micro finish. Constructed from durable tool steel, this gage is suitable for go inspection and for setting various measuring devices such as micrometers and verniers.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
Catalog No. 50-005-15096 Supplier MSC Supplier No. 88970504
This Class ZZ Plus plug gage is designed for precision inspection and calibration tasks. It features a 0.000200 in tolerance and a 10 micro finish. Constructed from durable tool steel, this gage is suitable for go inspection and for setting various measuring devices such as micrometers and verniers.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.