Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.77 mm Diameter, 2.000 in OAL
Catalog No. 5000512255
Encompass
This Class ZZ Plus plug gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. It offers high accuracy with a tight tolerance, ensuring reliable measurements for various industrial applications.
  • Overall length 2.000 in
  • Tolerance 0.00508 mm
  • Constructed from durable tool steel
  • Includes certificate of accuracy with NIST traceability
  • Suitable for go inspection and setting measuring devices
  • Features 10 micro coating/finish
  • Complies with ASME B89.1 standards
  • Diameter of measurement 0.77 mm
Catalog No. 50-005-12255 Supplier MSC Supplier No. 68091826
This Class ZZ Plus plug gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. It offers high accuracy with a tight tolerance, ensuring reliable measurements for various industrial applications.
  • Overall length 2.000 in
  • Tolerance 0.00508 mm
  • Constructed from durable tool steel
  • Includes certificate of accuracy with NIST traceability
  • Suitable for go inspection and setting measuring devices
  • Features 10 micro coating/finish
  • Complies with ASME B89.1 standards
  • Diameter of measurement 0.77 mm