Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.7780 in, 2.0000 in OAL
Catalog No. 5000530229
Encompass
This Class ZZ Plus Plug Gage is designed for high-precision measurement applications. Manufactured from durable tool steel, it features a 0.7780 inch diameter and a 2.0000 inch overall length, with a tight tolerance of 0.000200 inch and a smooth 10 micro finish. It is ideal for go inspection and setting various measuring devices.
  • 0.7780 inch diameter
  • 2.0000 inch overall length
  • 0.000200 inch tolerance
  • 10 micro finish
  • Made from tool steel
  • For go inspection
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
Catalog No. 50-005-30229 Supplier MSC Supplier No. 88977806
This Class ZZ Plus Plug Gage is designed for high-precision measurement applications. Manufactured from durable tool steel, it features a 0.7780 inch diameter and a 2.0000 inch overall length, with a tight tolerance of 0.000200 inch and a smooth 10 micro finish. It is ideal for go inspection and setting various measuring devices.
  • 0.7780 inch diameter
  • 2.0000 inch overall length
  • 0.000200 inch tolerance
  • 10 micro finish
  • Made from tool steel
  • For go inspection
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards