Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.78 mm Dia, Black Oxide Finish
Catalog No. 5000531028
Encompass
This plug gage is designed for precise "Go" inspection, as well as for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel with a black oxide finish, it provides reliable performance and accuracy in various industrial and laboratory settings.
  • Precise "Go" inspection
  • Used for setting micrometers, verniers, and other measuring devices
  • Made from durable tool steel
  • Features a black oxide finish
  • Includes a certificate of accuracy with NIST traceability standards ASME B89.1
  • Diameter of 0.78 mm
  • Overall length of 2.0000 inches
  • Tolerance of 0.00508 mm
Catalog No. 50-005-31028 Supplier MSC Supplier No. 72130644
This plug gage is designed for precise "Go" inspection, as well as for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel with a black oxide finish, it provides reliable performance and accuracy in various industrial and laboratory settings.
  • Precise "Go" inspection
  • Used for setting micrometers, verniers, and other measuring devices
  • Made from durable tool steel
  • Features a black oxide finish
  • Includes a certificate of accuracy with NIST traceability standards ASME B89.1
  • Diameter of 0.78 mm
  • Overall length of 2.0000 inches
  • Tolerance of 0.00508 mm