Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.7800 in Diameter, 2.0000 in Overall Length, Tool Steel
Catalog No. 5000530759
Encompass
This plug gage is a Class ZZ Plus tool, designed with a 0.7800 inch diameter and a 2.0000 inch overall length. Constructed from durable tool steel, it is suitable for precise go inspection and for setting various measuring devices.
  • Used for go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Constructed from tool steel for durability.
  • Features a 10 micro finish.
  • Offers a tolerance of 0.000200 "
Catalog No. 50-005-30759 Supplier MSC Supplier No. 88978002
This plug gage is a Class ZZ Plus tool, designed with a 0.7800 inch diameter and a 2.0000 inch overall length. Constructed from durable tool steel, it is suitable for precise go inspection and for setting various measuring devices.
  • Used for go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Constructed from tool steel for durability.
  • Features a 10 micro finish.
  • Offers a tolerance of 0.000200 "