Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.80 mm Diameter, 2.0000 in OAL
Catalog No. 5000531041
Encompass
This Class ZZ Plus Plug Gage is designed for precision "Go" inspection and for setting various measuring devices like micrometers and verniers. Made from durable tool steel, it offers accurate measurements with a 10 Micro finish and includes a Certificate of Accuracy with NIST Traceability.
  • Has a diameter of 0.80 mm.
  • Features a tolerance of 0.00508 mm.
  • Constructed from tool steel.
  • Designed for "Go" inspection and for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Overall length is 2.0000 inches.
  • Features a 10 Micro finish.
  • Meets ASME B89.1 standards.
Catalog No. 50-005-31041 Supplier MSC Supplier No. 68091859
This Class ZZ Plus Plug Gage is designed for precision "Go" inspection and for setting various measuring devices like micrometers and verniers. Made from durable tool steel, it offers accurate measurements with a 10 Micro finish and includes a Certificate of Accuracy with NIST Traceability.
  • Has a diameter of 0.80 mm.
  • Features a tolerance of 0.00508 mm.
  • Constructed from tool steel.
  • Designed for "Go" inspection and for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Overall length is 2.0000 inches.
  • Features a 10 Micro finish.
  • Meets ASME B89.1 standards.