Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.85 mm, Tool Steel, 10 Microfinish
Catalog No. 5000527892
Encompass
This Class ZZ Plus plug gage is designed for precise measurements and inspection. It is constructed from durable tool steel with a 10 microfinish, featuring a 0.85 mm diameter and an overall length of 2.0000 in. This gage is manufactured to ASME B89.1 standards, ensuring accuracy for various applications.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for go inspection applications.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Complies with ASME B89.1 standards.
  • Features a 10 microfinish.
  • Provides a precise tolerance of 0.00508 mm.
Catalog No. 50-005-27892 Supplier MSC Supplier No. 68091909
This Class ZZ Plus plug gage is designed for precise measurements and inspection. It is constructed from durable tool steel with a 10 microfinish, featuring a 0.85 mm diameter and an overall length of 2.0000 in. This gage is manufactured to ASME B89.1 standards, ensuring accuracy for various applications.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for go inspection applications.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Complies with ASME B89.1 standards.
  • Features a 10 microfinish.
  • Provides a precise tolerance of 0.00508 mm.