Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.8750 in Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000515401
Encompass
This plug gage is designed for Go inspection, and for setting micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Made from durable tool steel.
  • Manufactured to ASME B89.1 standards.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-15401 Supplier MSC Supplier No. 88987508
This plug gage is designed for Go inspection, and for setting micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Made from durable tool steel.
  • Manufactured to ASME B89.1 standards.
  • Features a 10 micro coating/finish.