Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.8950 in Diameter, 0.0002 in Tolerance, Steel
Catalog No. 5000530879
Encompass
This Class ZZ Plus Plug Gage is an economical general-purpose pin gage designed for quick and accurate measurements. It is used to measure hole sizes, calculate distances between holes, gage slot widths, and check locations. This gage provides added wear life and is suitable for go inspection, setting micrometers, verniers, and other measuring devices.
  • Class ZZ tolerance of 0.0002 in
  • Constructed from durable tool steel for extended wear life
  • Features a 10 microinch finish or better
  • Roundness maintained within 0.0001 in
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for go inspection
Catalog No. 50-005-30879 Supplier MSC Supplier No. 88989504
This Class ZZ Plus Plug Gage is an economical general-purpose pin gage designed for quick and accurate measurements. It is used to measure hole sizes, calculate distances between holes, gage slot widths, and check locations. This gage provides added wear life and is suitable for go inspection, setting micrometers, verniers, and other measuring devices.
  • Class ZZ tolerance of 0.0002 in
  • Constructed from durable tool steel for extended wear life
  • Features a 10 microinch finish or better
  • Roundness maintained within 0.0001 in
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for go inspection