Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 1.2958 in Diameter
Catalog No. 5000534800
Encompass
The Vermont Gage plug gage is a precision measurement tool designed for accurate verification of bore diameters. This Class ZZ Plus plug gage features a 1.2958 inch diameter and is suitable for various industrial and laboratory applications.
  • Class ZZ Plus for precise measurements.
  • 1.2958 inch diameter for specific sizing needs.
  • Handle is sold separately, allowing for customization or replacement.
Catalog No. 50-005-34800 Supplier MSC Supplier No. 64758691
The Vermont Gage plug gage is a precision measurement tool designed for accurate verification of bore diameters. This Class ZZ Plus plug gage features a 1.2958 inch diameter and is suitable for various industrial and laboratory applications.
  • Class ZZ Plus for precise measurements.
  • 1.2958 inch diameter for specific sizing needs.
  • Handle is sold separately, allowing for customization or replacement.