Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 1.3060 in Diameter
Catalog No. 5000528097
Encompass
Vermont Gage offers a Class ZZ Plus Plug Gage engineered for precise measurement. This plug gage features a 1.3060 inch diameter, ideal for verifying hole sizes and tolerances in various applications.
  • Offers Class ZZ Plus for high precision
  • Features a 1.3060 inch diameter of measurement
  • Handle sold separately, allowing for customizable setups
  • Made in the United States, ensuring quality craftsmanship
Catalog No. 50-005-28097 Supplier MSC Supplier No. 64759715
Vermont Gage offers a Class ZZ Plus Plug Gage engineered for precise measurement. This plug gage features a 1.3060 inch diameter, ideal for verifying hole sizes and tolerances in various applications.
  • Offers Class ZZ Plus for high precision
  • Features a 1.3060 inch diameter of measurement
  • Handle sold separately, allowing for customizable setups
  • Made in the United States, ensuring quality craftsmanship