Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 1.3078 in
Catalog No. 5000531032
Encompass
This plug gage is used for precisely measuring the diameter of holes or internal dimensions of parts. With a diameter of 1.3078 inches, it offers high accuracy for quality control and manufacturing processes. It is designed for checking the tolerance of drilled or machined holes.
  • Precisely measures internal diameters.
  • High accuracy for quality control applications.
  • Ideal for checking tolerance of drilled or machined holes.
  • Handle is sold separately.
Catalog No. 50-005-31032 Supplier MSC Supplier No. 64759897
This plug gage is used for precisely measuring the diameter of holes or internal dimensions of parts. With a diameter of 1.3078 inches, it offers high accuracy for quality control and manufacturing processes. It is designed for checking the tolerance of drilled or machined holes.
  • Precisely measures internal diameters.
  • High accuracy for quality control applications.
  • Ideal for checking tolerance of drilled or machined holes.
  • Handle is sold separately.