Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 1.39 mm, Tool Steel, Black Oxide
Catalog No. 5000528298
Encompass
This Class ZZ Plus Plug Gage has a diameter of 1.39 mm and is made from durable tool steel with a black oxide finish. It features a tolerance of 0.00508 mm and an overall length of 2.0000 inches, making it suitable for precise measurement tasks.
  • Tolerance of 0.00508 mm
  • Overall length of 2.0000 inches
  • Made from tool steel
  • Black oxide finish
  • Used for Go inspection; set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards
Catalog No. 50-005-28298 Supplier MSC Supplier No. 50795152
This Class ZZ Plus Plug Gage has a diameter of 1.39 mm and is made from durable tool steel with a black oxide finish. It features a tolerance of 0.00508 mm and an overall length of 2.0000 inches, making it suitable for precise measurement tasks.
  • Tolerance of 0.00508 mm
  • Overall length of 2.0000 inches
  • Made from tool steel
  • Black oxide finish
  • Used for Go inspection; set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards