Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 1.59 mm, 2.0000 in, Tool Steel
Catalog No. 5000511004
Encompass
This precision plug gage is crafted from tool steel and features a 1.59 mm diameter with an overall length of 2.0000 inches. It is designed for 'Go' inspection and setting various measuring devices, ensuring accurate measurements in critical applications.
  • Constructed from durable tool steel
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
  • Features a 10 microfinish
  • Maintains a tolerance of 0.00508 mm
Catalog No. 50-005-11004 Supplier MSC Supplier No. 68092642
This precision plug gage is crafted from tool steel and features a 1.59 mm diameter with an overall length of 2.0000 inches. It is designed for 'Go' inspection and setting various measuring devices, ensuring accurate measurements in critical applications.
  • Constructed from durable tool steel
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
  • Features a 10 microfinish
  • Maintains a tolerance of 0.00508 mm