Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 1.98 mm Diameter, Tool Steel
Catalog No. 5000530637
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. Featuring a 1.98 mm diameter and a 2.0000 in overall length, it offers a fine 0.00508 mm tolerance. Crafted from durable tool steel with a 10 micro coating/finish, it ensures reliability and accuracy. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Designed for Go inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Meets ASME B89.1 standards.
  • Manufactured from high-quality tool steel.
  • Provides a 1.98 mm measurement diameter.
  • Features a 0.00508 mm tolerance.
  • Has a 2.0000 in overall length.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Finished with a 10 micro coating.
Catalog No. 50-005-30637 Supplier MSC Supplier No. 68093038
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. Featuring a 1.98 mm diameter and a 2.0000 in overall length, it offers a fine 0.00508 mm tolerance. Crafted from durable tool steel with a 10 micro coating/finish, it ensures reliability and accuracy. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Designed for Go inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Meets ASME B89.1 standards.
  • Manufactured from high-quality tool steel.
  • Provides a 1.98 mm measurement diameter.
  • Features a 0.00508 mm tolerance.
  • Has a 2.0000 in overall length.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Finished with a 10 micro coating.