Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 12.38 mm, 2.0000 in OAL
Catalog No. 5000512673
Encompass
This Class ZZ Plus Plug Gage is designed for high-precision measurement applications. It features a 12.38 mm diameter and a 2.0000 inch overall length, crafted from durable Tool Steel. This gage is ideal for Go inspection and setting various measuring devices like micrometers and verniers, ensuring accuracy and reliability in demanding environments. It adheres to ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability.
  • Overall length of 2.0000 inches
  • Tolerance of 0.00508 mm
  • Made from tool steel
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards
  • Features a 10 micro coating/finish
  • Diameter of 12.38 mm
Catalog No. 50-005-12673 Supplier MSC Supplier No. 68103431
This Class ZZ Plus Plug Gage is designed for high-precision measurement applications. It features a 12.38 mm diameter and a 2.0000 inch overall length, crafted from durable Tool Steel. This gage is ideal for Go inspection and setting various measuring devices like micrometers and verniers, ensuring accuracy and reliability in demanding environments. It adheres to ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability.
  • Overall length of 2.0000 inches
  • Tolerance of 0.00508 mm
  • Made from tool steel
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards
  • Features a 10 micro coating/finish
  • Diameter of 12.38 mm