Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 12.98 mm Diameter, Tool Steel
Catalog No. 5000531350
Encompass
This Class ZZ Plus plug gage is a high-precision tool for accurate 'Go' inspection and setting measuring devices like micrometers and verniers. Made from durable tool steel, it features a 12.98 mm diameter, 2.0000-inch overall length, and a tight 0.00508 mm tolerance. It includes a NIST traceable Certificate of Accuracy and meets ASME B89.1 standards, offering reliable performance with a 10 micro coating/finish.
  • Designed for accurate 'Go' inspection.
  • Sets micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Features a 12.98 mm diameter.
  • Offers a tight 0.00508 mm tolerance.
  • Includes NIST traceable certificate of accuracy.
  • Complies with ASME B89.1 standards.
  • 10 micro coating/finish.
Catalog No. 50-005-31350 Supplier MSC Supplier No. 68104033
This Class ZZ Plus plug gage is a high-precision tool for accurate 'Go' inspection and setting measuring devices like micrometers and verniers. Made from durable tool steel, it features a 12.98 mm diameter, 2.0000-inch overall length, and a tight 0.00508 mm tolerance. It includes a NIST traceable Certificate of Accuracy and meets ASME B89.1 standards, offering reliable performance with a 10 micro coating/finish.
  • Designed for accurate 'Go' inspection.
  • Sets micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Features a 12.98 mm diameter.
  • Offers a tight 0.00508 mm tolerance.
  • Includes NIST traceable certificate of accuracy.
  • Complies with ASME B89.1 standards.
  • 10 micro coating/finish.