Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 13.01 mm, Tool Steel
Catalog No. 5000536295
Encompass
This plug gage is engineered for precise 'Go Inspection' and for calibrating various measuring devices such as micrometers and verniers. Constructed from durable tool steel, it ensures reliable performance and accuracy in metrology applications.
  • Designed for go inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Made from durable tool steel.
  • Features a 10 micro finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Has a diameter of 13.01 mm.
  • Overall length is 2.0000 inches.
Catalog No. 50-005-36295 Supplier MSC Supplier No. 68104066
This plug gage is engineered for precise 'Go Inspection' and for calibrating various measuring devices such as micrometers and verniers. Constructed from durable tool steel, it ensures reliable performance and accuracy in metrology applications.
  • Designed for go inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Made from durable tool steel.
  • Features a 10 micro finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Has a diameter of 13.01 mm.
  • Overall length is 2.0000 inches.