Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 13.15 mm, 2.0000 in OAL
Catalog No. 5000528920
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and calibration in various applications. It is made from durable tool steel and meets ASME B89.1 standards, ensuring reliable performance for quality control and instrument calibration.
  • Designed for go inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from tool steel for durability
  • Features a 10 micro coating/finish
  • Adheres to ASME B89.1 standards
  • Has a diameter of 13.15 mm
  • Measures 2.0000 inches in overall length
  • Provides a tolerance of 0.00508 mm
Catalog No. 50-005-28920 Supplier MSC Supplier No. 68104207
This Class ZZ Plus Plug Gage is designed for precise measurement and calibration in various applications. It is made from durable tool steel and meets ASME B89.1 standards, ensuring reliable performance for quality control and instrument calibration.
  • Designed for go inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from tool steel for durability
  • Features a 10 micro coating/finish
  • Adheres to ASME B89.1 standards
  • Has a diameter of 13.15 mm
  • Measures 2.0000 inches in overall length
  • Provides a tolerance of 0.00508 mm