Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 13.41 mm Diameter, Tool Steel
Catalog No. 5000527963
Encompass
This Class ZZ Plus Plug Gage is a precision measuring instrument with a 13.41 mm diameter, constructed from durable tool steel. It is designed for accurate 'Go' inspection and is suitable for setting various measuring devices such as micrometers and verniers. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance.
  • Class ZZ Plus Plug Gage
  • 13.41 mm diameter
  • Made from tool steel
  • For Go inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Features a 10 micro coating/finish
  • Tolerance of 0.00508 mm
Catalog No. 50-005-27963 Supplier MSC Supplier No. 68104462
This Class ZZ Plus Plug Gage is a precision measuring instrument with a 13.41 mm diameter, constructed from durable tool steel. It is designed for accurate 'Go' inspection and is suitable for setting various measuring devices such as micrometers and verniers. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance.
  • Class ZZ Plus Plug Gage
  • 13.41 mm diameter
  • Made from tool steel
  • For Go inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Features a 10 micro coating/finish
  • Tolerance of 0.00508 mm