Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 13.70 mm Dia, 2.0000 in OAL
Catalog No. 5000527297
Encompass
This Class ZZ Plus plug gage is designed for precise "Go" inspection and setting micrometers, verniers, and other measuring devices. It features a 13.70 mm diameter and a 2.0000 inch overall length, with a tolerance of 0.00508 mm.
  • Made from tool steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Features a 10 micro finish.
Catalog No. 50-005-27297 Supplier MSC Supplier No. 68104751
This Class ZZ Plus plug gage is designed for precise "Go" inspection and setting micrometers, verniers, and other measuring devices. It features a 13.70 mm diameter and a 2.0000 inch overall length, with a tolerance of 0.00508 mm.
  • Made from tool steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Features a 10 micro finish.