Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 13.99 mm, 2 in., Steel
Catalog No. 5000532038
Encompass
The product is a Class ZZ Plus Plug Gage, constructed from tool steel with a 10 Micro Finish. It features a 13.99 mm diameter and a 2 in. overall length, providing a tolerance of 0.005mm. This gage is ideal for Go inspection and for setting micrometers, verniers, and other measuring devices, ensuring precision in measurement applications. It includes a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards.
  • Class ZZ Plus gauge for precise measurements
  • Constructed from durable tool steel
  • Features a 10 Micro Finish for accuracy
  • Suitable for Go inspection applications
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards
Catalog No. 50-005-32038 Supplier MSC Supplier No. 68105048
The product is a Class ZZ Plus Plug Gage, constructed from tool steel with a 10 Micro Finish. It features a 13.99 mm diameter and a 2 in. overall length, providing a tolerance of 0.005mm. This gage is ideal for Go inspection and for setting micrometers, verniers, and other measuring devices, ensuring precision in measurement applications. It includes a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards.
  • Class ZZ Plus gauge for precise measurements
  • Constructed from durable tool steel
  • Features a 10 Micro Finish for accuracy
  • Suitable for Go inspection applications
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards