Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 17.33 mm Diameter, Tool Steel, ASME B89.1
Catalog No. 5000530980
Encompass
The product is a Class ZZ Plus Plug Gage, featuring a 17.33 mm diameter and an overall length of 2.0000 inches. It has a tolerance of 0.00508 mm and a 10 Micro coating/finish. It is made of Tool Steel and is used for Go Inspection, setting micrometers, verniers, and other measuring devices.
  • Made from tool steel.
  • Overall length of 2.0000 inches.
  • Tolerance of 0.00508 mm.
  • Diameter of 17.33 mm.
  • Features a 10 micro coating/finish.
  • Conforms to ASME B89.1 standards.
  • Suitable for go inspection and setting various measuring devices.
Catalog No. 50-005-30980 Supplier MSC Supplier No. 68108380
The product is a Class ZZ Plus Plug Gage, featuring a 17.33 mm diameter and an overall length of 2.0000 inches. It has a tolerance of 0.00508 mm and a 10 Micro coating/finish. It is made of Tool Steel and is used for Go Inspection, setting micrometers, verniers, and other measuring devices.
  • Made from tool steel.
  • Overall length of 2.0000 inches.
  • Tolerance of 0.00508 mm.
  • Diameter of 17.33 mm.
  • Features a 10 micro coating/finish.
  • Conforms to ASME B89.1 standards.
  • Suitable for go inspection and setting various measuring devices.