Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 17.61 mm, 0.00508 mm Tolerance
Catalog No. 5000512952
Encompass
This Class ZZ Plus plug gage from Vermont Gage is designed for precise measurement applications. It features a diameter of 17.61 mm with a tight tolerance of 0.00508 mm and an overall length of 2.0000 inches.
  • Factory warranty.
  • New condition.
  • Free shipping.
  • Authorized distributor.
Catalog No. 50-005-12952 Supplier MSC Supplier No. 68108661
This Class ZZ Plus plug gage from Vermont Gage is designed for precise measurement applications. It features a diameter of 17.61 mm with a tight tolerance of 0.00508 mm and an overall length of 2.0000 inches.
  • Factory warranty.
  • New condition.
  • Free shipping.
  • Authorized distributor.