Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 18.27 mm, 2.0000 in OAL
Catalog No. 5000528924
Encompass
This plug gage is designed for precise measurement and inspection, featuring a diameter of 18.27 mm and an overall length of 2.0000 inches. Crafted from tool steel with a 10 micro coating/finish, it is suitable for 'Go' inspection and setting various measuring devices.
  • For Go inspection
  • Set micrometers, verniers, and other measuring devices
  • Crafted from tool steel
  • Features a 10 micro coating/finish
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Country of origin: United States
Catalog No. 50-005-28924 Supplier MSC Supplier No. 68109321
This plug gage is designed for precise measurement and inspection, featuring a diameter of 18.27 mm and an overall length of 2.0000 inches. Crafted from tool steel with a 10 micro coating/finish, it is suitable for 'Go' inspection and setting various measuring devices.
  • For Go inspection
  • Set micrometers, verniers, and other measuring devices
  • Crafted from tool steel
  • Features a 10 micro coating/finish
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Country of origin: United States