Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 18.77 mm, Steel, 2.0000 in
Catalog No. 5000528038
Encompass
This precision plug gage is designed for accurate "Go" inspection, checking hole sizes, and setting other measuring devices. It features a 18.77 mm diameter and a 2.0000 inch overall length. Constructed from tool steel with a 10 microinch finish, it conforms to ASME B89.1 standards and includes a Certificate of Accuracy with NIST traceability.
  • Designed for accurate "Go" inspection
  • Checks hole sizes and sets other measuring devices
  • Made from durable tool steel
  • Features a 10 microinch finish for precision
  • Conforms to ASME B89.1 standards
  • Includes a Certificate of Accuracy with NIST traceability for reliable measurements
Catalog No. 50-005-28038 Supplier MSC Supplier No. 68109826
This precision plug gage is designed for accurate "Go" inspection, checking hole sizes, and setting other measuring devices. It features a 18.77 mm diameter and a 2.0000 inch overall length. Constructed from tool steel with a 10 microinch finish, it conforms to ASME B89.1 standards and includes a Certificate of Accuracy with NIST traceability.
  • Designed for accurate "Go" inspection
  • Checks hole sizes and sets other measuring devices
  • Made from durable tool steel
  • Features a 10 microinch finish for precision
  • Conforms to ASME B89.1 standards
  • Includes a Certificate of Accuracy with NIST traceability for reliable measurements