Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 19.39 mm, Tool Steel
Supplier:  MSC 68110444
Encompass
This Vermont Gage Class ZZ Plus plug gage features a 19.39 mm diameter and is constructed from durable tool steel. It is designed for precise 'Go' inspection, setting micrometers, verniers, and other measuring instruments. Adhering to ASME B89.1 standards, this gage ensures accuracy and reliability for critical measurements, making it suitable for a variety of laboratory and industrial applications. Each gage includes a Certificate of Accuracy with NIST traceability, confirming its precision.
  • Designed for Go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards
  • Constructed from tool steel
  • Tolerance of 0.00508 mm
  • 10 micro coating/finish
  • 2.000 in overall length
  • 19.39 mm diameter of measurement
Catalog No. 50-005-27966