Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 19.63 mm Dia, 2.0000 in OAL
Catalog No. 5000528925
Encompass
This Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection of bore sizes. Made from durable tool steel, it ensures reliable performance for quality control applications.
  • 19.63 mm diameter
  • 2.0000 in overall length
  • 0.00508 mm tolerance
  • 10 Micro surface finish
  • Durable tool steel construction
  • Suitable for Go Inspection applications
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Includes Certificate of Accuracy with NIST Traceability
  • Adheres to ASME B89.1 standards
Catalog No. 50-005-28925 Supplier MSC Supplier No. 68110683
This Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection of bore sizes. Made from durable tool steel, it ensures reliable performance for quality control applications.
  • 19.63 mm diameter
  • 2.0000 in overall length
  • 0.00508 mm tolerance
  • 10 Micro surface finish
  • Durable tool steel construction
  • Suitable for Go Inspection applications
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Includes Certificate of Accuracy with NIST Traceability
  • Adheres to ASME B89.1 standards