Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 2.29 mm Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000530357
Encompass
This precision plug gage is designed for 'go' inspection and for setting micrometers, verniers, and other measuring devices. Manufactured from durable tool steel, it features a 10 micro finish and includes a Certificate of Accuracy with NIST Traceability to ensure reliable measurements.
  • Designed for go inspection
  • Can set micrometers, verniers, and other measuring devices
  • Constructed from tool steel
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Features a 10 micro finish
  • Has a diameter of 2.29 mm
  • Offers an overall length of 2.0000 in
  • Provides a tolerance of 0.00508 mm
Catalog No. 50-005-30357 Supplier MSC Supplier No. 68093343
This precision plug gage is designed for 'go' inspection and for setting micrometers, verniers, and other measuring devices. Manufactured from durable tool steel, it features a 10 micro finish and includes a Certificate of Accuracy with NIST Traceability to ensure reliable measurements.
  • Designed for go inspection
  • Can set micrometers, verniers, and other measuring devices
  • Constructed from tool steel
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Features a 10 micro finish
  • Has a diameter of 2.29 mm
  • Offers an overall length of 2.0000 in
  • Provides a tolerance of 0.00508 mm