Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 2.59 mm, Tool Steel
Catalog No. 5000527913
Encompass
Vermont Gage Class ZZ Plus Plug Gage, featuring a 2.59 mm diameter and a 2.0000 in overall length. It is constructed from durable tool steel, designed for 'Go' inspection, and is also suitable for setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy, traceable to NIST Standards ASME B89.1.
  • Class ZZ Plus tolerance
  • 2.59 mm diameter
  • 2.0000 in overall length
  • Constructed from tool steel
  • Designed for 'Go' inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy
  • NIST Traceability Standards ASME B89.1
Catalog No. 50-005-27913 Supplier MSC Supplier No. 68093640
Vermont Gage Class ZZ Plus Plug Gage, featuring a 2.59 mm diameter and a 2.0000 in overall length. It is constructed from durable tool steel, designed for 'Go' inspection, and is also suitable for setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy, traceable to NIST Standards ASME B89.1.
  • Class ZZ Plus tolerance
  • 2.59 mm diameter
  • 2.0000 in overall length
  • Constructed from tool steel
  • Designed for 'Go' inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy
  • NIST Traceability Standards ASME B89.1