Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 20.60 mm Diameter, Tool Steel
Catalog No. 5000528040
Encompass
This Vermont Gage Class ZZ Plus Plug Gage features a 20.60 mm diameter, crafted from durable tool steel for precision measurement tasks. It is designed to provide accurate and reliable dimensional checks in various industrial applications.
  • Class ZZ Plus design
  • 20.60 mm diameter for precise measurement
  • Durable tool steel construction
  • Handle sold separately
Catalog No. 50-005-28040 Supplier MSC Supplier No. 68111657
This Vermont Gage Class ZZ Plus Plug Gage features a 20.60 mm diameter, crafted from durable tool steel for precision measurement tasks. It is designed to provide accurate and reliable dimensional checks in various industrial applications.
  • Class ZZ Plus design
  • 20.60 mm diameter for precise measurement
  • Durable tool steel construction
  • Handle sold separately