Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 21.00 mm diameter, 2.0000 in OAL, 0.00508 mm tolerance
Catalog No. 5000527979
Encompass
This plug gage is designed for precise 'Go' inspection and for setting micrometers, verniers, and other measuring devices. It is constructed from tool steel and adheres to ASME B89.1 standards, ensuring high accuracy and reliability.
  • Certificate of accuracy with NIST traceability
  • Constructed from durable tool steel
  • Ideal for 'Go' inspection and setting measuring devices
  • Conforms to ASME B89.1 standards
  • Features a 10 micro coating/finish
Catalog No. 50-005-27979 Supplier MSC Supplier No. 68112051
This plug gage is designed for precise 'Go' inspection and for setting micrometers, verniers, and other measuring devices. It is constructed from tool steel and adheres to ASME B89.1 standards, ensuring high accuracy and reliability.
  • Certificate of accuracy with NIST traceability
  • Constructed from durable tool steel
  • Ideal for 'Go' inspection and setting measuring devices
  • Conforms to ASME B89.1 standards
  • Features a 10 micro coating/finish