Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 21.70 mm, 0.00508 mm Tolerance
Catalog No. 5000527441
Encompass
This plug gage is designed for precise measurement applications. It features a Class ZZ Plus classification, ensuring high accuracy for critical dimensional checks. With a 21.70 mm diameter and a strict tolerance of 0.00508 mm, it is suitable for a variety of industrial and laboratory needs.
  • Class ZZ Plus for high accuracy
  • 21.70 mm diameter
  • 2.0000 in overall length
  • 0.00508 mm tolerance for critical measurements
  • 10 microinch finish
Catalog No. 50-005-27441 Supplier MSC Supplier No. 68112754
This plug gage is designed for precise measurement applications. It features a Class ZZ Plus classification, ensuring high accuracy for critical dimensional checks. With a 21.70 mm diameter and a strict tolerance of 0.00508 mm, it is suitable for a variety of industrial and laboratory needs.
  • Class ZZ Plus for high accuracy
  • 21.70 mm diameter
  • 2.0000 in overall length
  • 0.00508 mm tolerance for critical measurements
  • 10 microinch finish